4-Methylthiazole-5-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

M1812-1G M1812-5G
TCM1812-1GEA 49.11 CAD
TCM1812-1G TCM1812-5G
4-Methylthiazole-5-carbaldehyde ≥98.0% (by GC)
4-Methylthiazole-5-carbaldehyde

CAS Number: 82294-70-0
MDL Number: MFCD07368277
Molecular Formula: C5H5NOS
Molecular Weight: 127.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 76


Other Product Names / Synonyms: 5-Formyl-4-methylthiazole

Formula: C₅H₅NOS
Boiling Pt: 118 °C
Melting Pt: 76 °C
MDL Number: MFCD07368277
CAS Number: 82294-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d269fc28fccba31be6ac6cf7d5190ea5cdf30045&vid=d269fc28fccba31be6ac6cf7d5190ea5cdf30045&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR