4-Methoxyphenyl-3-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M1725-1G M1725-5G
TCM1725-1GEA 234.79 CAD
TCM1725-1G TCM1725-5G
4-Methoxyphenyl-3-O-benzyl-β-D-galactopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-benzyl-beta-D-galactopyranoside

CAS Number: 383905-60-0
Molecular Formula: C20H24O7
Molecular Weight: 376.41
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 154
Storage Temperature: 0-10°C

Formula: C₂₀H₂₄O₇
Melting Pt: 154 °C
CAS Number: 383905-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f1434e147aab09554f376ac3a43b7d9e9d8cb51&vid=9f1434e147aab09554f376ac3a43b7d9e9d8cb51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR