Gal[26Bn]β(1-4)Glc[236Bn]-β-MP ≥98.0%
Supplier: TCI America
Synonyms:
LacMP 3'4'-OH, per OBn, 4-Methoxyphenyl 4-O-(2,6-Di-O-benzyl-β-D-galactopyranosyl)-2,3,6-tri-O-benzyl-β-D-glucopyranoside
CAS Number: 358681-61-5
Molecular Formula: C54H58O12
Molecular Weight: 899.05
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: 4 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Glycosylation, Glycosylation, Bioscience, Disaccharides, Glycosides, p-Methoxyphenyl Glycosides, O-Substituted Sugars
Other Product Names / Synonyms: 4-Methoxyphenyl 4-O-(2,6-Di-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside, LacMP 3'4'-OH, per OBn
Formula:
C₅₄H₅₈O₁₂ MW: 899.05 g/mol Melting Pt: 130 °C |
CAS Number:
358681-61-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...