4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

M1604-1G M1604-5G
TCM1604-1GEA 226.41 CAD
TCM1604-1G TCM1604-5G
4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-3-O-allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside

Molecular Weight: 545.59
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Molecular Formula: C31H31NO8

Formula: C₃₁H₃₁NO₈
CAS Number: 1820583-64-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR