4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) radical_free
Supplier: TCI America
M1531-1G
M1531-5G
TCM1531-1GEA
173.8
CAD
TCM1531-1G
TCM1531-5G
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) radical_free
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 15051-46-4
MDL Number: MFCD17677357
Molecular Formula: C13H22NO3
Molecular Weight: 240.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 89
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Monomers, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer
Formula:
C₁₃H₂₂NO₃ Melting Pt: 89 °C |
MDL Number:
MFCD17677357 CAS Number: 15051-46-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...