1-Methyl-1H-indole-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Indole-2-carboxylic acid, 1-methyl-, 1-Methylindole-2-carboxylic acid

M1519-1G M1519-5G
TCM1519-1GEA 79 CAD
TCM1519-1G TCM1519-5G
1-Methyl-1H-indole-2-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methyl-1H-indole-2-carboxylic acid

CAS Number: 16136-58-6
MDL Number: MFCD00005801
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 212

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00005801
CAS Number: 16136-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1add9a1d1dd30b3d491cf6ad5973344f5f96fa1c&vid=1add9a1d1dd30b3d491cf6ad5973344f5f96fa1c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR