4-Morpholinoaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-(4-Morpholinyl)aniline

M1466-1G M1466-5G
TCM1466-1GEA 37.92 CAD
TCM1466-1G TCM1466-5G
4-Morpholinoaniline ≥98.0% (by GC, titration analysis)
4-Morpholinoaniline

CAS Number: 2524-67-6
MDL Number: MFCD00006169
Molecular Formula: C10H14N2O
Molecular Weight: 178.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 4-(4-Aminophenyl)morpholine

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00006169
CAS Number: 2524-67-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5334e61c71fdc3fe62414409adf2dfd305824e87&vid=5334e61c71fdc3fe62414409adf2dfd305824e87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR