Bis(2-mercaptoethyl)sulfide ≥97.0% (by GC)

Supplier: TCI America

M1337-250G M1337-25G
TCM1337-250GEA 1363.54 CAD
TCM1337-250G TCM1337-25G
Bis(2-mercaptoethyl)sulfide ≥97.0% (by GC)
Bis(2-mercaptoethyl)sulfide

CAS Number: 3570-55-6
MDL Number: MFCD00004893
Molecular Formula: C4H10S3
Molecular Weight: 154.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 90
Freezing point (°C): -11
Specific Gravity (20/20): 1.19


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-Mercaptoethyl Sulfide, 2,2'-Thiodiethanethiol

Formula: C₄H₁₀S₃
Boiling Pt: 138 °C
Density: 1.19 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00004893
CAS Number: 3570-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a78f3f3e47df0b549ce8fecf06465517f244de2&vid=9a78f3f3e47df0b549ce8fecf06465517f244de2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR