(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥98.0%
Supplier: TCI America
Synonyms:
(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid
[Optical Resolving]
CAS Number: 17257-71-5
MDL Number: MFCD00064200
Molecular Formula: C10H9F3O3
Molecular Weight: 234.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 34
Specific rotation [a]20/D: -72 deg (C=2, MeOH)
Product Categories: Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (-)-Mosher's Acid, (-)-MTPA
Formula:
C₁₀H₉F₃O₃ MW: 234.17 g/mol Flash Pt: 113 °C |
MDL Number:
MFCD00064200 CAS Number: 17257-71-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...