2-Methyl-2-nitropropane-1,3-diol ≥98.0% (by total nitrogen basis)

Supplier: TCI America

M0639-25G M0639-5G
TCM0639-25GEA 398.16 CAD
TCM0639-25G TCM0639-5G
2-Methyl-2-nitropropane-1,3-diol ≥98.0% (by total nitrogen basis)
2-Methyl-2-nitropropane-1,3-diol

CAS Number: 77-49-6
MDL Number: MFCD00024812
Molecular Formula: C4H9NO4
Molecular Weight: 135.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 156

Formula: C₄H₉NO₄
Melting Pt: 156 °C
MDL Number: MFCD00024812
CAS Number: 77-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ebd668e9ea6d55a7ac017e4cce0e4dc9e9f300&vid=25ebd668e9ea6d55a7ac017e4cce0e4dc9e9f300&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR