Ethyl methanesulfonate ≥99.0% (by GC)
Supplier: TCI America
M0607-25G
M0607-100G
TCM0607-25GEA
110.35
CAD
TCM0607-25G
TCM0607-100G
Ethyl methanesulfonate ≥99.0% (by GC)
Ethyl methanesulphonate
CAS Number: 62-50-0
MDL Number: MFCD00007559
Molecular Formula: C3H8O3S
Molecular Weight: 124.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 100
Specific Gravity (20/20): 1.21
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Methanesulfonic Acid Ethyl Ester
Formula:
C₃H₈O₃S Boiling Pt: 89 °C Density: 1.21 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00007559 CAS Number: 62-50-0 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...