(-)-Menthoxyacetic acid

Supplier: TCI America

Synonyms: 2-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)acetic acid, (-)-Menthyloxyacetic acid

M0573-25G M0573-5G
TCM0573-25GEA 951.16 CAD
TCM0573-25G TCM0573-5G
(-)-Menthoxyacetic acid
(-)-Menthoxyacetic Acid

CAS Number: 40248-63-3
MDL Number: MFCD00001483
Molecular Formula: C12H22O3
Molecular Weight: 214.31
Form: Crystal
Color: Very Pale Yellow
Flash Point (°C): 110
Specific rotation [a]20/D: -94 deg (C=2, EtOH)

Formula: C₁₂H₂₂O₃
MW: 214.3 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00001483
CAS Number: 40248-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3133be67c6905d1b7a9eaab9390311ca46a2e32f&vid=3133be67c6905d1b7a9eaab9390311ca46a2e32f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR