Loganin ≥98.0% (by HPLC)

Supplier: TCI America

L0268-10MG L0268-50MG
TCL0268-10MGEA 120.71 CAD
TCL0268-10MG TCL0268-50MG
Loganin ≥98.0% (by HPLC)
Loganin

CAS Number: 18524-94-2
MDL Number: MFCD00075645
Molecular Formula: C17H26O10
Molecular Weight: 390.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 225
Specific rotation [a]20/D: -85 deg (C=0.1, MeOH)
Lambda max.: 240 nm (MeOH)

Formula: C₁₇H₂₆O₁₀
Melting Pt: 225 °C
MDL Number: MFCD00075645
CAS Number: 18524-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64d0e937afbdb8a276807f6ef40802f8c6d0d6da&vid=64d0e937afbdb8a276807f6ef40802f8c6d0d6da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR