Limonin ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: (4aS,6aR,8aR,8bR,9aS,12S,12aS,14aR,14bR)-12-(Furan-3-yl)-6,6,8a,12a-tetramethyldecahydrooxireno[2,3-d]pyrano[4',3':3,3a]isobenzofuro[5,4-f]isochromene-3,8,10(1H,6H,8aH)-trione

L0258-100MG L0258-1G
TCL0258-100MGEA 69.52 CAD
TCL0258-100MG TCL0258-1G
Limonin ≥95.0% (by HPLC)
Limonin

CAS Number: 1180-71-8
MDL Number: MFCD00075922
Molecular Formula: C26H30O8
Molecular Weight: 470.52
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 298
Specific rotation [a]20/D: -117 deg (C=1, Acetone)
Lambda max.: 285 nmStorage Temperature: 0-10°C

Formula: C₂₆H₃₀O₈
MW: 470.52 g/mol
Melting Pt: 298 °C
MDL Number: MFCD00075922
CAS Number: 1180-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6faadaa911d16bce5661cddba9c09cfd5b6c73c&vid=c6faadaa911d16bce5661cddba9c09cfd5b6c73c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR