Lumefantrine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

L0256-25G L0256-5G
TCL0256-25GEA 172.66 CAD
TCL0256-25G TCL0256-5G
Lumefantrine ≥98.0% (by HPLC, titration analysis)
Lumefantrine

CAS Number: 82186-77-4
MDL Number: MFCD05662268
Molecular Formula: C30H32Cl3NO
Molecular Weight: 528.94
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 136


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients, Antiprotozoal Ingredients
Other Product Names / Synonyms: Benflumetol, (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol

Formula: C₃₀H₃₂Cl₃NO
Melting Pt: 136 °C
MDL Number: MFCD05662268
CAS Number: 82186-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d348452b280f062716c57cda65bdd2099349be7&vid=5d348452b280f062716c57cda65bdd2099349be7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR