Lithium dodecyl sulfate ≥98.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulfate lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulfate lithium salt, Dodecyl sulfuric acid lithium salt, Lithium lauryl sulfate, Lauryl sulphate lithium salt

L0254-25G L0254-5G
TCL0254-25GEA 268.6 CAD
TCL0254-25G TCL0254-5G
Lithium dodecyl sulfate ≥98.0% (by titrimetric analysis) for biochemical research
Lithium dodecyl sulphate

[for Biochemical Research]
CAS Number: 2044-56-6
MDL Number: MFCD00007467
Molecular Formula: C12H26O4S
Molecular Weight: 272.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Anionic Surfactants, Bioscience, Anionic Surfactants (for Biochemistry)
Other Product Names / Synonyms: Dodecylsulfuric Acid Lithium Salt, Lithium Lauryl Sulfate, Laurylsulfuric Acid Lithium Salt, LDS

Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
MDL Number: MFCD00007467
CAS Number: 2044-56-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db4f6c0cb980174d8c42c3e2da92a798f47414a9&vid=5c88527c39a9655f819863d6dca61f01016e745f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR