Lomerizine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

L0245-1G L0245-200MG
TCL0245-1GEA 457.88 CAD
TCL0245-1G TCL0245-200MG
Lomerizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Lomerizine dihydrochloride

CAS Number: 101477-54-7
MDL Number: MFCD01703867
Molecular Formula: C27H30F2N2O3
Molecular Weight: 541.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 1-[Bis(4-fluorophenyl)methyl]-4-(2,3,4-trimethoxybenzyl)piperazine Dihydrochloride

Formula: C₂₇H₃₀F₂N₂O₃
Melting Pt: 180 °C
MDL Number: MFCD01703867
CAS Number: 101477-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb95c327a28875a134641c652d2c0000288a2de&vid=ceb95c327a28875a134641c652d2c0000288a2de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR