D-(-)-Leucinol ≥97.0%

Supplier: TCI America

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

L0236-1G L0236-5G
TCL0236-1GEA 42.66 CAD
TCL0236-1G TCL0236-5G
D-(-)-Leucinol ≥97.0%
D(-)-Leucinol

CAS Number: 53448-09-2
MDL Number: MFCD00004734
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 90
Specific Gravity (20/20): 0.91
Specific rotation [a]20/D: -3 deg (C=9, EtOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-2-Amino-4-methyl-1-pentanol

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199
Density: 0.91 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00004734
CAS Number: 53448-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce3d7f380650d1169dbc28aed9d453d60266ed8d&vid=ce3d7f380650d1169dbc28aed9d453d60266ed8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR