Lanthanum(III) oxide ≥99.9%

Supplier: TCI America

L0235-25G L0235-500G
TCL0235-25GEA 71.1 CAD
TCL0235-25G TCL0235-500G
Lanthanum(III) oxide ≥99.9%
Lanthanum (III) oxide

CAS Number: 1312-81-8
MDL Number: MFCD00011071
Molecular Formula: La2O3
Molecular Weight: 325.81
Form: Crystal
Color: White

Formula: La₂O₃
MDL Number: MFCD00011071
CAS Number: 1312-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c74e7931509d6b19df31e8cd763e7c1fcef77f78&vid=c74e7931509d6b19df31e8cd763e7c1fcef77f78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR