Lac-β(1-4)lac-β-C10 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Decyl 4-O-β-D-Lactopyranosyl-β-D-lactopyranoside

L0229-100MG L0229-10MG
TCL0229-100MGEA 1429.74 CAD
TCL0229-100MG TCL0229-10MG
Lac-β(1-4)lac-β-C10 ≥98.0% (by HPLC)
Lac-beta(1-4)lac-beta-C10

Molecular Weight: 806.85
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C
Molecular Formula: C34H62O21


Other Product Names / Synonyms: Decyl 4-O-beta-D-Lactopyranosyl-beta-D-lactopyranoside

Formula: C₃₄H₆₂O₂₁
MW: 806.85 g/mol
CAS Number: 1858224-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b4d416debae7571e526669668a78b29db90e40&vid=c1b4d416debae7571e526669668a78b29db90e40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR