L-Leucine-tert-butyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, H-Leu-OtBu·HCl, L-leucine 1,1-dimethylethyl ester hydrochloride, H-Leu-OtBu.HCl, TERT-BUTYL (2S)-2-AMINO-4-METHYLPENTANOATE HYDROCHLORIDE

L0197-1G L0197-5G
TCL0197-1GEA 37.92 CAD
TCL0197-1G TCL0197-5G
L-Leucine-tert-butyl ester hydrochloride ≥98.0%
L-Leucine-tert-butyl ester hydrochloride

CAS Number: 2748-02-9
MDL Number: MFCD00038906
Molecular Formula: C10H21NO2
Molecular Weight: 223.74
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: 20 deg (C=2, EtOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Leu-OtBu.HCl

Formula: C₁₀H₂₁NO₂
Melting Pt: 167 °C
MDL Number: MFCD00038906
CAS Number: 2748-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eed9ec26a3c23b28cb02f7f83b51f5350a9e387&vid=3eed9ec26a3c23b28cb02f7f83b51f5350a9e387&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR