Kojic acid dipalmitate ≥98.0% (by HPLC)

Supplier: TCI America

K0050-25G K0050-5G
TCK0050-25GEA 115.66 CAD
TCK0050-25G TCK0050-5G
Kojic acid dipalmitate ≥98.0% (by HPLC)
Kojic acid dipalmitate

CAS Number: 79725-98-7
MDL Number: MFCD03840587
Molecular Formula: C38H66O6
Molecular Weight: 618.94
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 96


Other Product Names / Synonyms: 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one

Formula: C₃₈H₆₆O₆
Melting Pt: 96 °C
MDL Number: MFCD03840587
CAS Number: 79725-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4940d826dc420e31f18776de76cb11d4e337f03f&vid=4940d826dc420e31f18776de76cb11d4e337f03f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR