Isoquinoline-5-carbaldehyde ≥98.0%

Supplier: TCI America

I0942-1G I0942-5G
TCI0942-1GEA 246.48 CAD
TCI0942-1G TCI0942-5G
Isoquinoline-5-carbaldehyde ≥98.0%
Isoquinoline-5-carbaldehyde

CAS Number: 80278-67-7
MDL Number: MFCD03412483
Molecular Formula: C10H7NO
Molecular Weight: 157.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119

Formula: C₁₀H₇NO
Melting Pt: 119 °C
MDL Number: MFCD03412483
CAS Number: 80278-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc400f95d3cf96fcb9a911db609f5a43e8d0e87f&vid=fc400f95d3cf96fcb9a911db609f5a43e8d0e87f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR