4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

I0941-1G I0941-5G
TCI0941-1GEA 93.22 CAD
TCI0941-1G TCI0941-5G
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC, titration analysis)
4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine

CAS Number: 151266-23-8
MDL Number: MFCD03787931
Molecular Formula: C5H4IN5
Molecular Weight: 261.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Lambda max.: 282 nm


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 4-Amino-3-iodo-1H-pyrazolo[3,4-d]pyrimidine

Formula: C₅H₄N₅I
MW: 261.03 g/mol
MDL Number: MFCD03787931
CAS Number: 151266-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2329c4b7b1fe460150efaeeaf8def193becf5e90&vid=2329c4b7b1fe460150efaeeaf8def193becf5e90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR