Isobutylamine hydroiodide ≥97.0%

Supplier: TCI America

I0935-1G I0935-5G
TCI0935-1GEA 133.83 CAD
TCI0935-1G TCI0935-5G
Isobutylamine hydroiodide ≥97.0%
Isobutylamine hydroiodide

CAS Number: 205508-75-4
Molecular Formula: C4H11N
Molecular Weight: 201.05
Purity/Analysis Method: >97.0% (N,T)
Form: Crystal

Formula: C₄H₁₁N
MW: 201.05 g/mol
CAS Number: 205508-75-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60bc081e5d71cec9b0d69f158d96acb5f2eed19e&vid=60bc081e5d71cec9b0d69f158d96acb5f2eed19e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR