(1-Indolyl)acetic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Indole-1-acetic Acid, 2-(1H-Indol-1-yl)acetic acid
I0932-1G
I0932-5G
TCI0932-1GEA
74.26
CAD
TCI0932-1G
TCI0932-5G
(1-Indolyl)acetic acid ≥97.0% (by titrimetric analysis)
(1-Indolyl)acetic acid
CAS Number: 24297-59-4
MDL Number: MFCD00047262
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 170
Other Product Names / Synonyms: Indole-1-acetic Acid
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 170 °C |
MDL Number:
MFCD00047262 CAS Number: 24297-59-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...