5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde, 7-Isopropyl-1,4-dimethylazulene-3-carboxaldehyde

I0928-1G I0928-200MG
TCI0928-1GEA 794.74 CAD
TCI0928-1G TCI0928-200MG
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde ≥97.0%
5-Isopropyl-3,8-dimethylazulene-1-carbaldehyde

CAS Number: 3331-47-3
MDL Number: MFCD00275723
Molecular Formula: C16H18O
Molecular Weight: 226.32
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 83


Other Product Names / Synonyms: 5-Isopropyl-3,8-dimethylazulene-1-carboxaldehyde

Formula: C₁₆H₁₈O
MW: 226.32
Melting Pt: 83 °C
MDL Number: MFCD00275723
CAS Number: 3331-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48708a62651e38ed1198a1db835b801324266605&vid=48708a62651e38ed1198a1db835b801324266605&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR