5-Iodo-o-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
I0916-25G
I0916-5G
TCI0916-25GEA
87.53
CAD
TCI0916-25G
TCI0916-5G
5-Iodo-o-toluic acid ≥98.0% (by GC, titration analysis)
5-Iodo-o-toluic acid
CAS Number: 54811-38-0
MDL Number: MFCD01570258
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 181
Other Product Names / Synonyms: 5-Iodo-2-methylbenzoic Acid
Formula:
C₈H₇IO₂ Melting Pt: 181 °C |
MDL Number:
MFCD01570258 CAS Number: 54811-38-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...