3-Iodo-9-phenylcarbazole ≥98.0% (by GC)
Supplier: TCI America
I0913-1G
I0913-5G
TCI0913-1GEA
58.46
CAD
TCI0913-1G
TCI0913-5G
3-Iodo-9-phenylcarbazole ≥98.0% (by GC)
3-Iodo-9-phenylcarbazole
CAS Number: 502161-03-7
MDL Number: MFCD09261276
Molecular Formula: C18H12IN
Molecular Weight: 369.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 106
Formula:
C₁₈H₁₂IN Melting Pt: 106 °C |
MDL Number:
MFCD09261276 CAS Number: 502161-03-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...