Isopentyl hexanoate ≥98.0%

Supplier: TCI America

Synonyms: Isopentyl hexanoate, Isoamyl caproate, Isoamyl hexanoate, Hexanoic acid Isopentyl ester, Hexanoic acid isoamyl ester, Isopentyl caproate, Caproic acid isoamyl ester, Caproic acid isopentyl ester, 3-Methylbutyl hexanoate, Hexanoic acid isopentyl ester

I0899-100ML I0899-25ML
TCI0899-100MLEA 115.34 CAD
TCI0899-100ML TCI0899-25ML
Isopentyl hexanoate ≥98.0%
Isopentyl hexanoate

CAS Number: 2198-61-0
MDL Number: MFCD00027280
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 88
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Hexanoic Acid Isoamyl Ester, Isopentyl Hexanoate, Hexanoic Acid Isopentyl Ester

Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 226
Density: 0.86 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00027280
CAS Number: 2198-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd985a9e054af7aea1915cdadaf7abcaad19e6df&vid=fd985a9e054af7aea1915cdadaf7abcaad19e6df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR