3-Isopropylphenylboronic acid 97.0-113.0% (contains varying amounts of Anhydride)
Supplier: TCI America
I0887-1G
I0887-5G
TCI0887-1GEA
238.58
CAD
TCI0887-1G
TCI0887-5G
3-Isopropylphenylboronic acid 97.0-113.0% (contains varying amounts of Anhydride)
3-Isopropylphenylboronic acid
CAS Number: 216019-28-2
MDL Number: MFCD01074634
Molecular Formula: C9H13BO2
Molecular Weight: 164.01
Form: Crystal
Color: White
Flash Point (°C): 110
Other Product Names / Synonyms: 3-Isopropylbenzeneboronic Acid
Formula:
C₉H₁₃BO₂ Flash Pt: 110 °C |
MDL Number:
MFCD01074634 CAS Number: 216019-28-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...