2-Iodo-4-nitroaniline ≥98.0% (by GC)

Supplier: TCI America

I0878-1G I0878-5G
TCI0878-1GEA 77.74 CAD
TCI0878-1G TCI0878-5G
2-Iodo-4-nitroaniline ≥98.0% (by GC)
2-Iodo-4-nitroaniline

CAS Number: 6293-83-0
MDL Number: MFCD00955758
Molecular Formula: C6H5IN2O2
Molecular Weight: 264.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107
Lambda max.: 360 nm (MeOH)

Formula: C₆H₅IN₂O₂
Melting Pt: 107 °C
MDL Number: MFCD00955758
CAS Number: 6293-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4077286540b64737ef4d8a849aa35520a4d082b8&vid=4077286540b64737ef4d8a849aa35520a4d082b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR