N-Isopropylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

I0877-1G I0877-5G
TCI0877-1GEA 96.38 CAD
TCI0877-1G TCI0877-5G
N-Isopropylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
N-Isopropylhydroxylamine hydrochloride

CAS Number: 50632-53-6
MDL Number: MFCD00012599
Molecular Formula: C3H9NO
Molecular Weight: 111.57
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 72
Storage Temperature: <0°C

Formula: C₃H₉NO
Melting Pt: 72 °C
MDL Number: MFCD00012599
CAS Number: 50632-53-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb7b326c89658456ef3a9e0d356c18e4d1c946c8&vid=bb7b326c89658456ef3a9e0d356c18e4d1c946c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR