2-Iodo-9,9-dimethylfluorene ≥98.0% (by GC)

Supplier: TCI America

I0875-25G I0875-5G
TCI0875-25GEA 680.98 CAD
TCI0875-25G TCI0875-5G
2-Iodo-9,9-dimethylfluorene ≥98.0% (by GC)
2-Iodo-9,9-dimethylfluorene

CAS Number: 144981-85-1
MDL Number: MFCD12024272
Molecular Formula: C15H13I
Molecular Weight: 320.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 66

Formula: C₁₅H₁₃I
Melting Pt: 66 °C
MDL Number: MFCD12024272
CAS Number: 144981-85-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f9031c35f1bf61bbe9dfca282b403a52314da87&vid=9f9031c35f1bf61bbe9dfca282b403a52314da87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR