3-Iodobenzyl alcohol ≥98.0% (by GC)
Supplier: TCI America
I0866-25G
I0866-5G
TCI0866-25GEA
309.68
CAD
TCI0866-25G
TCI0866-5G
3-Iodobenzyl alcohol ≥98.0% (by GC)
3-Iodobenzyl alcohol
CAS Number: 57455-06-8
MDL Number: MFCD00004635
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 113
Specific Gravity (20/20): 1.84
Formula:
C₇H₇IO Boiling Pt: 105 °C Density: 1.84 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00004635 CAS Number: 57455-06-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...