2-Iodobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

I0858-25G I0858-5G
TCI0858-25GEA 282.66 CAD
TCI0858-25G TCI0858-5G
2-Iodobenzyl alcohol ≥98.0% (by GC)
2-Iodobenzyl alcohol

CAS Number: 5159-41-1
MDL Number: MFCD00004610
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 92

Formula: C₇H₇IO
Boiling Pt: 145 °C
Melting Pt: 92 °C
MDL Number: MFCD00004610
CAS Number: 5159-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b866abe7e58fd6896b71b8aaa2de28f2355ea6f7&vid=b866abe7e58fd6896b71b8aaa2de28f2355ea6f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR