Indole-2-carbaldehyd ≥97.0%

Supplier: TCI America

Synonyms: Indole-2-carboxaldehyde, 1H-Indole-2-carbaldehyde

I0852-1G I0852-5G
TCI0852-1GEA 186.44 CAD
TCI0852-1G TCI0852-5G
Indole-2-carbaldehyd ≥97.0%
Indole-2-carbaldehyd

CAS Number: 19005-93-7
MDL Number: MFCD03001425
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 139


Other Product Names / Synonyms: 2-Formylindole

Formula: C₉H₇NO
MW: 145.16
Melting Pt: 139 °C
MDL Number: MFCD03001425
CAS Number: 19005-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36525e59a72ff7c92595e91253b82000f853fc16&vid=36525e59a72ff7c92595e91253b82000f853fc16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR