3-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0851-1G I0851-5G
TCI0851-1GEA 108.61 CAD
TCI0851-1G TCI0851-5G
3-Formylbenzoic acid ≥98.0% (by HPLC, titration analysis)
3-Formylbenzoic acid

CAS Number: 619-21-6
MDL Number: MFCD00039575
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 177


Other Product Names / Synonyms: 3-Carboxybenzaldehyde, 3-Formylbenzoic Acid

Formula: C₈H₆O₃
Melting Pt: 177 °C
MDL Number: MFCD00039575
CAS Number: 619-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd32d906b96ba91041b2e35df7572cc53eca52fb&vid=fd32d906b96ba91041b2e35df7572cc53eca52fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR