Idebenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0848-25G I0848-5G
TCI0848-25GEA 824.76 CAD
TCI0848-25G TCI0848-5G
Idebenone ≥98.0% (by HPLC, titration analysis)
Idebenone

CAS Number: 58186-27-9
MDL Number: MFCD00274552
Molecular Formula: C19H30O5
Molecular Weight: 338.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 53


Product Categories: Bioscience, Antioxidants, Vitamin Related Compounds
Other Product Names / Synonyms: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone

Formula: C₁₉H₃₀O₅
Melting Pt: 53 °C
MDL Number: MFCD00274552
CAS Number: 58186-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fca859b9f7ae8b09a57ae539c3fa4419c033036&vid=8fca859b9f7ae8b09a57ae539c3fa4419c033036&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR