Ivabradine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
(S)-3-(3-(((3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methyl)(methyl)amino)propyl)-7,8-dimethoxy-1,3,4,5-tetrahydro-2H-benzo[d]azepin-2-one hydrochloride, Ivabradine HCl
I0847-1G
I0847-200MG
TCI0847-1GEA
1158.14
CAD
TCI0847-1G
TCI0847-200MG
Ivabradine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ivabradine hydrochloride
CAS Number: 148849-67-6
MDL Number: MFCD00929899
Molecular Formula: C27H36N2O5
Molecular Weight: 505.05
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: 7 deg (C=1, DMSO)
Storage Temperature: 0-10°C
Formula:
C₂₇H₃₆N₂O₅ MW: 505.05 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00929899 CAS Number: 148849-67-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...