4-Iodophenol ≥98.0% (by GC) for biochemical research

Supplier: TCI America

I0840-1G I0840-5G
TCI0840-1GEA 41.08 CAD
TCI0840-1G TCI0840-5G
4-Iodophenol ≥98.0% (by GC) for biochemical research
4-Iodophenol

[for Biochemical Research]
CAS Number: 540-38-5
MDL Number: MFCD00002327
Molecular Formula: C6H5IO
Molecular Weight: 220.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93

Formula: C₆H₅IO
MW: 220.01 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00002327
CAS Number: 540-38-5
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b481c6b5d4fd61bc54d9c752441e08ac9a00a31&vid=9083e9bd60106986f62d013e0845eca2ba86e040&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR