4-Iodo-1-methyl-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

I0829-1G I0829-5G
TCI0829-1GEA 67.94 CAD
TCI0829-1G TCI0829-5G
4-Iodo-1-methyl-1H-pyrazole ≥98.0% (by GC)
4-Iodo-1-methyl-1H-pyrazole

CAS Number: 39806-90-1
MDL Number: MFCD05663860
Molecular Formula: C4H5IN2
Molecular Weight: 208.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64

Formula: C₄H₅IN₂
Melting Pt: 64 °C
MDL Number: MFCD05663860
CAS Number: 39806-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f9a7d83b2415dc3209615ba7fdf9744fffc258c&vid=0f9a7d83b2415dc3209615ba7fdf9744fffc258c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR