L-Isoserine 98,0

Supplier: TCI America

Synonyms: (S)-3-Amino-2-hydroxypropanoic acid, L-Isoserine, (S)-3-Amino-2-hydroxypropionic Acid

I0828-1G I0828-5G
TCI0828-1GEA 36.34 CAD
TCI0828-1G TCI0828-5G
L-Isoserine 98,0
L-Isoserine

CAS Number: 632-13-3
MDL Number: MFCD03428326
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 197
Specific rotation [a]20/D: -33 deg (C=1, H2O)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-3-Amino-2-hydroxypropionic Acid

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 197 °C
MDL Number: MFCD03428326
CAS Number: 632-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a906fabe951ff6a24ffba4db62db3baa0ba7de9&vid=0a906fabe951ff6a24ffba4db62db3baa0ba7de9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR