Isopropyl acetoacetate ≥97.0%

Supplier: TCI America

Synonyms: Isopropyl 3-oxobutanoate

I0826-25G I0826-500G
TCI0826-25GEA 83.57 CAD
TCI0826-25G TCI0826-500G
Isopropyl acetoacetate ≥97.0%
Isopropyl acetoacetate

CAS Number: 542-08-5
MDL Number: MFCD00043933
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 71
Specific Gravity (20/20): 0.99


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Acetoacetic Acid Isopropyl Ester, Isopropyl 3-Oxobutyrate, 3-Oxobutyric Acid Isopropyl Ester

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 187 °C
Density: 0.99 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00043933
CAS Number: 542-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a837c4a88915d90e8cddf48339ee4cbf292e1ee5&vid=a837c4a88915d90e8cddf48339ee4cbf292e1ee5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR