1-Isocyanoadamantane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Adamantyl Isocyanide

I0824-1G I0824-5G
TCI0824-1GEA 243.32 CAD
TCI0824-1G TCI0824-5G
1-Isocyanoadamantane ≥97.0% (by GC)
1-Isocyanoadamantane

CAS Number: 22110-53-8
MDL Number: MFCD02258889
Molecular Formula: C11H15N
Molecular Weight: 161.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 197


Product Categories: Synthetic Organic Chemistry, Isonitriles (Multi-component Reaction)
Other Product Names / Synonyms: 1-Adamantyl Isocyanide

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Melting Pt: 197 °C
MDL Number: MFCD02258889
CAS Number: 22110-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=372bc6a6d095b6b0d271212766e08c36bcf4a87b&vid=372bc6a6d095b6b0d271212766e08c36bcf4a87b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR