Isoliquiritigenin ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
I0822-100MG
I0822-1G
TCI0822-100MGEA
44.24
CAD
TCI0822-100MG
TCI0822-1G
Isoliquiritigenin ≥97.0% (by HPLC, titration analysis)
Isoliquiritigenin
CAS Number: 961-29-5
MDL Number: MFCD00075907
Molecular Formula: C15H12O4
Molecular Weight: 256.26
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198
Storage Temperature: 0-10°C
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: (E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one, 2',4,4'-Trihydroxychalcone
Formula:
C₁₅H₁₂O₄ Melting Pt: 198 °C |
MDL Number:
MFCD00075907 CAS Number: 961-29-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...