4-Imidazol-1-ylbenzaldehyde ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(1-Imidazolyl)benzaldehyde, 1-(4-Formylphenyl)imidazole, 4-(1H-Imidazol-1-yl)benzaldehyde
I0818-1G
I0818-5G
TCI0818-1GEA
44.24
CAD
TCI0818-1G
TCI0818-5G
4-Imidazol-1-ylbenzaldehyde ≥97.0% (by GC, titration analysis)
4-Imidazol-1-ylbenzaldehyde
CAS Number: 10040-98-9
MDL Number: MFCD00209977
Molecular Formula: C10H8N2O
Molecular Weight: 172.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 149
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 1-(4-Formylphenyl)imidazole
Formula:
C₁₀H₈N₂O MW: 172.1833 g/mol Melting Pt: 149 °C |
MDL Number:
MFCD00209977 CAS Number: 10040-98-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...