Isopropyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: Isopropylparaben, Isopropyl parahydroxybenzoate, Isopropyl p-Hydroxybenzoate, 4-Hydroxybenzoic acid isopropyl ester, Isopropyl Paraben, Isopropyl 4-Hydroxybenzoate

I0817-25G I0817-5G
TCI0817-25GEA 65.92 CAD
TCI0817-25G TCI0817-5G
Isopropyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research
Isopropyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 4191-73-5
MDL Number: MFCD00016468
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >99.0% (T)
Form: Powder
Boiling point (°C): 160
Melting point (°C): 86


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Isopropyl Ester, Isopropylparaben

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 160 °C
Melting Pt: 86 °C
MDL Number: MFCD00016468
CAS Number: 4191-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=425ad6a8ae4b3284e742b36e408871d15cfb6713&vid=a9ba9e0b01ef7dab3ae4d88de6fb5cdcd93769f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR