Isobutyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

I0816-25G I0816-5G
TCI0816-25GEA 102.57 CAD
TCI0816-25G TCI0816-5G
Isobutyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research
Isobutyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 4247-02-3
MDL Number: MFCD00020167
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 76


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Isobutyl Ester, Isobutylparaben

Formula: C₁₁H₁₄O₃
Melting Pt: 76 °C
MDL Number: MFCD00020167
CAS Number: 4247-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80dabe8eceddade9322e1acc1048bae0520e73af&vid=dbbf4566bc542613ec835a418d29349d4d2d5a05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR