Isoamyl 4-methoxycinnamate ≥95.0%
Supplier: TCI America
Synonyms:
4-Methoxycinnamic Acid Isoamyl Ester, 4-Methoxycinnamic Acid Isopentyl Ester, Amiloxate, Isopentyl 4-Methoxycinnamate, Isopentyl 3-(4-methoxyphenyl)acrylate
CAS Number: 71617-10-2
MDL Number: MFCD00583856
Molecular Formula: C15H20O3
Molecular Weight: 248.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 1.04
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 4-Methoxycinnamic Acid Isoamyl Ester, Isopentyl 4-Methoxycinnamate, 4-Methoxycinnamic Acid Isopentyl Ester
Formula:
C₁₅H₂₀O₃ MW: 248.32 g/mol Boiling Pt: 158 °C Density: 1.04 (20 °C) |
MDL Number:
MFCD00583856 CAS Number: 71617-10-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...